siRNA Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000553.4 | NP_000544.2 | 6 | 1422 | ||
XM_011544639.2 | XP_011542941.1 | 6 | 904 | |||
GenBank mRNA | AF091214.1 | - | 6 | 865 | ||
AY818673.1 | - | 5 | 634 |
