siRNA Details
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005433.3 | NP_005424.1 | 9, 10 | 1368 | ||
| XM_005258139.3 | XP_005258196.1 | 9, 10 | 1330 | |||
| XM_017025960.1 | XP_016881449.1 | 9, 10 | 1210 | |||
| GenBank mRNA | AK314063.1 | - | 9, 10 | 1312 | ||
| BC048960.1 | - | 9, 10 | 1364 | 5260751 | ||
| M15990.1 | - | 9, 10 | 1354 | |||

YES1
Genomic Map