siRNA Details
| Gene Symbol: ZIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003412.3 | NP_003403.2 | 3 | 1947 | ||
| GenBank mRNA | BC104848.1 | - | 3 | 1301 | 8143851 | |
| BC104850.1 | - | 3 | 1302 | 8143853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZIC1| Gene Symbol: ZIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003412.3 | NP_003403.2 | 3 | 1947 | ||
| GenBank mRNA | BC104848.1 | - | 3 | 1301 | 8143851 | |
| BC104850.1 | - | 3 | 1302 | 8143853 | ||