siRNA Details
| Gene Symbol: ZNF708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 660 | ||
| NM_001297561.1 | NP_001284490.1 | 3 | 451 | |||
| NM_021269.2 | NP_067092.2 | 4 | 576 | |||
| XM_017027201.1 | XP_016882690.1 | 4 | 478 | |||
| XM_017027202.1 | XP_016882691.1 | 4 | 601 | |||
| XM_017027203.1 | XP_016882692.1 | 2 | 381 | |||
| XM_017027204.1 | XP_016882693.1 | 4 | 393 | |||
| GenBank mRNA | AK128184.1 | - | 4 | 535 | ||
| CD704852.1 | - | 3 | 473 | |||
| DB216212.1 | - | 3 | 449 | |||

ZNF708
Genomic Map