siRNA Details
| Gene Symbol: ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001203250.1 | NP_001190179.1 | 4 | 1101 | ||
| NM_021143.3 | NP_066966.2 | 4 | 1110 | |||
| GenBank mRNA | AK023094.1 | - | 4 | 1004 | ||
| AL080125.1 | - | 4 | 982 | |||
| BC036714.2 | - | 4 | 1028 | 4846514 | ||
| BC045814.1 | - | 4 | 945 | 4830061 | ||
| Gene Symbol: ZNF625-ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037802.1 | 8 | 1467 | |||
| GenBank mRNA | AK096560.1 | - | 8 | 1467 | ||

ZNF20
Genomic Map