siRNA Details
| Gene Symbol: ZNF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006963.4 | NP_008894.2 | 2 | 801 | ||
| GenBank mRNA | AK291508.1 | - | 2 | 703 | ||
| AK312278.1 | - | 2 | 703 | |||
| BC010642.1 | - | 2 | 691 | 3852749 | ||
| BC026193.1 | - | 7 | 769 | |||
| BC041139.1 | - | 2 | 1152 | 5751460 | ||
| BC053687.1 | - | 2 | 695 | 5214077 | ||
| BT009806.1 | - | 1 | 558 | |||
| S50223.1 | - | 1 | 647 | |||
| X52346.1 | - | 1 | 144 | |||

ZNF22
Genomic Map