siRNA Details
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267033.1 | NP_001253962.1 | 3 | 1115 | ||
| NM_003422.2 | NP_003413.2 | 3 | 797 | |||
| NM_198055.1 | NP_932172.1 | 3 | 1115 | |||
| XM_005259204.3 | XP_005259261.1 | 4 | 1470 | |||
| XM_011527264.2 | XP_011525566.1 | 3 | 1199 | |||
| XM_017027206.1 | XP_016882695.1 | 3 | 1300 | |||
| GenBank mRNA | AB209481.1 | - | 1 | 1176 | ||
| AF055077.1 | - | 3 | 797 | |||
| AF055078.1 | - | 3 | 1115 | |||
| AK308893.1 | - | 2 | 1248 | |||
| AK310381.1 | - | 3 | 822 | |||
| BC007777.2 | - | 3 | 773 | 4301723 | ||
| BC053316.1 | - | 3 | 838 | |||

MZF1
Genomic Map