siRNA Details
| Gene Symbol: RELL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085399.1 | NP_001078868.1 | 2 | 321 | ||
| NM_001085400.1 | NP_001078869.1 | 2 | 321 | |||
| GenBank mRNA | AK075468.1 | - | 2 | 320 | ||
| BC039540.1 | - | 2 | 319 | 5748847 | ||
| DA400382.1 | - | 2 | 321 | |||

RELL1
Genomic Map