siRNA Details
| Gene Symbol: ZNF174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032292.2 | NP_001027463.1 | 1, 2 | 1216 | ||
| NM_003450.2 | NP_003441.1 | 1, 2 | 1216 | |||
| GenBank mRNA | AF542096.1 | - | 1, 2 | 432 | ||
| BC000876.1 | - | 1, 2 | 1143 | 3461658 | ||
| BC001161.1 | - | 1, 2 | 1162 | 3533665 | ||
| BT006923.1 | - | 1, 2 | 401 | |||
| U31248.1 | - | 1, 2 | 986 | |||

ZNF174
Genomic Map