siRNA Details
| Gene Symbol: ZNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006526.2 | NP_006517.1 | 1 | 586 | ||
| XM_005260545.3 | XP_005260602.1 | 2 | 816 | |||
| XM_006723875.2 | XP_006723938.1 | 4 | 997 | |||
| XM_011529036.1 | XP_011527338.1 | 4 | 959 | |||
| XM_017028059.1 | XP_016883548.1 | 2 | 741 | |||
| GenBank mRNA | AF041259.1 | - | 1 | 586 | ||
| AK290350.1 | - | 2 | 813 | |||
| BC113427.1 | - | 1 | 414 | 8322479 | ||

ZNF217
Genomic Map