siRNA Details
| Gene Symbol: ZNF236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306089.1 | NP_001293018.1 | 23 | 4146 | ||
| NM_007345.3 | NP_031371.3 | 23 | 4337 | |||
| XM_005266762.3 | XP_005266819.1 | 23 | 4489 | |||
| XM_011526165.2 | XP_011524467.1 | 24 | 4491 | |||
| XM_011526166.2 | XP_011524468.1 | 22 | 3963 | |||
| XM_011526168.2 | XP_011524470.1 | 23 | 4203 | |||
| XM_011526169.2 | XP_011524471.1 | 23 | 4202 | |||
| GenBank mRNA | AB208834.1 | - | 15 | 2765 | ||
| AF085243.1 | - | 23 | 4337 | |||
| AF085244.1 | - | 23 | 4337 | |||
| BC140859.1 | - | 23 | 4168 | 9021727 | ||
| BX648028.1 | - | 23 | 4146 | |||

ZNF236
Genomic Map