siRNA Details
| Gene Symbol: RAB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004637.5 | NP_004628.4 | 5, 6 | 768 | ||
| GenBank mRNA | AF498942.1 | - | 4, 5 | 536 | ||
| AK094449.1 | - | 4, 5 | 449 | |||
| AK225177.1 | - | 4, 5 | 522 | |||
| AK225691.1 | - | 4, 5 | 515 | |||
| AK290721.1 | - | 5, 6 | 672 | |||
| AK298344.1 | - | 5, 6 | 649 | |||
| AK308934.1 | - | 5, 6 | 570 | |||
| BC008721.2 | - | 5, 6 | 711 | 2821435 | ||
| BC013728.1 | - | 4, 5 | 551 | 3882701 | ||
| U44104.1 | - | 4, 5 | 536 | |||

RAB7A
Genomic Map