siRNA Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000387.5 | NP_000378.1 | 5, 6 | 744 | ||
| GenBank mRNA | AK301744.1 | - | 6, 7 | 705 | ||
| AK312962.1 | - | 5, 6 | 637 | |||
| BC001689.2 | - | 5, 6 | 616 | 3050073 | ||
| BI913588.1 | - | 5, 6 | 647 | |||
| Y10319.1 | - | 5, 6 | 607 | |||

SLC25A20
Genomic Map