siRNA Details
| Gene Symbol: TMEM109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024092.2 | NP_076997.1 | 2 | 480 | ||
| GenBank mRNA | AK303961.1 | - | 2 | 426 | ||
| AK313913.1 | - | 2 | 264 | |||
| AX775849.1 | - | 2 | 152 | |||
| AX775851.1 | - | 2 | 174 | |||
| BC001309.1 | - | 2 | 174 | 3453616 | ||
| BI463800.1 | - | 2 | 511 | |||

TMEM109
Genomic Map