siRNA Details
| Gene Symbol: TMEM109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024092.2 | NP_076997.1 | 3, 4 | 720 | ||
| GenBank mRNA | AK303961.1 | - | 3, 4 | 666 | ||
| AK313913.1 | - | 3, 4 | 504 | |||
| AX775849.1 | - | 3, 4 | 392 | |||
| AX775851.1 | - | 3, 4 | 414 | |||
| BC001309.1 | - | 3, 4 | 414 | 3453616 | ||

TMEM109
Genomic Map