siRNA Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301011.1 | NP_001287940.1 | 4 | 853 | ||
| NM_024296.4 | NP_077272.2 | 4 | 853 | |||
| XM_011542113.2 | XP_011540415.1 | 4 | 1612 | |||
| XM_017002306.1 | XP_016857795.1 | 4 | 853 | |||
| XM_017002307.1 | XP_016857796.1 | 4 | 1607 | |||
| GenBank mRNA | AK225113.1 | - | 4 | 623 | ||
| AK291904.1 | - | 4 | 863 | |||
| BC022848.2 | - | 4 | 587 | 3946778 | ||
| BM981930.1 | - | 3 | 254 | |||

CCDC28B
Genomic Map