siRNA Details
| Gene Symbol: CENPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199803.2 | NP_001186732.1 | 6 | 1192 | ||
| NM_001322101.1 | NP_001309030.1 | 7 | 1306 | |||
| NM_024322.3 | NP_077298.1 | 7 | 1325 | |||
| NR_136182.1 | 6 | 1110 | ||||
| NR_136183.1 | 6 | 1211 | ||||
| NR_136184.1 | 6 | 1091 | ||||
| GenBank mRNA | AK027859.1 | - | 6 | 926 | ||
| AK315483.1 | - | 7 | 951 | |||
| AL514456.3 | - | 6 | 884 | |||
| BC002870.1 | - | 7 | 982 | 3942844 | ||
| CX872478.1 | - | 6 | 728 | |||

CENPO
Genomic Map