siRNA Details
| Gene Symbol: ZNF576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145347.1 | NP_001138819.1 | 3 | 540 | ||
| NM_024327.2 | NP_077303.1 | 3 | 576 | |||
| GenBank mRNA | AK026353.1 | - | 3 | 576 | ||
| BC002981.1 | - | 3 | 501 | 3535991 | ||
| BP358040.1 | - | 3 | 540 | |||

ZNF576
Genomic Map