siRNA Details
| Gene Symbol: THOC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142350.1 | NP_001135822.1 | 2 | 415 | ||
| NM_024339.3 | NP_077315.2 | 2 | 415 | |||
| GenBank mRNA | AK075330.1 | - | 2 | 382 | ||
| AK308960.1 | - | 2 | 403 | |||
| AK310113.1 | - | 2 | 415 | |||
| AK314086.1 | - | 2 | 393 | |||
| AL832548.1 | - | 1 | 19 | |||
| BC003118.1 | - | 3 | 295 | 3537243 | ||
| BC050674.1 | - | 2 | 354 | 6067702 | ||

THOC6
Genomic Map