siRNA Details
| Gene Symbol: LRRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024512.4 | NP_078788.2 | 2 | 410 | ||
| XM_011534110.2 | XP_011532412.1 | 2 | 199 | |||
| XM_017007177.1 | XP_016862666.1 | 2 | 410 | |||
| GenBank mRNA | AJ308569.2 | - | 2 | 184 | ||
| AK057808.1 | - | 2 | 252 | |||
| AK223440.1 | - | 2 | 252 | |||
| AK314658.1 | - | 2 | 173 | |||
| AK315636.1 | - | 2 | 193 | |||
| BC029118.1 | - | 2 | 182 | 5183639 | ||
| DA144445.1 | - | 2 | 410 | |||

LRRC2
Genomic Map