siRNA Details
| Gene Symbol: WDR25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161476.1 | NP_001154948.1 | 2 | 767 | ||
| NM_024515.4 | NP_078791.3 | 2 | 898 | |||
| XM_005268056.4 | XP_005268113.1 | 2 | 774 | |||
| XM_011537150.2 | XP_011535452.1 | 2 | 1021 | |||
| GenBank mRNA | AY295082.1 | - | 2 | 781 | ||
| BX248767.1 | - | 2 | 760 | |||

WDR25
Genomic Map