siRNA Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024539.3 | NP_078815.3 | 6 | 1180 | ||
| NM_194463.1 | NP_919445.1 | 6 | 1343 | |||
| GenBank mRNA | AK027169.1 | - | 4 | 352 | ||
| AK074264.1 | - | 6 | 1343 | |||
| AK126553.1 | - | 6 | 1163 | |||
| BC030951.1 | - | 5 | 456 | |||
| BC036901.1 | - | 6 | 1192 | |||
| BC056677.1 | - | 4 | 327 | |||
| BC063404.1 | - | 6 | 1182 | 5191785 | ||

RNF128
Genomic Map