siRNA Details
| Gene Symbol: TANGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024562.1 | NP_078838.1 | 11 | 1974 | ||
| XM_011523327.2 | XP_011521629.1 | 11 | 2033 | |||
| GenBank mRNA | AB051533.1 | - | 11 | 1974 | ||
| AK022750.1 | - | 5 | 638 | |||
| AK095333.1 | - | 10 | 1520 | |||

TANGO6
Genomic Map