siRNA Details
| Gene Symbol: SNRNP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024571.3 | NP_078847.1 | 2 | 255 | ||
| GenBank mRNA | AK026593.1 | - | 2 | 224 | ||
| BC001381.1 | - | 2 | 1077 | 3051476 | ||
| BC009179.1 | - | 2 | 206 | 3946937 | ||
| Z84714.1 | - | 2 | 169 | |||

SNRNP25
Genomic Map