siRNA Details
| Gene Symbol: ARHGAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024605.3 | NP_078881.3 | 3, 4 | 310 | ||
| XM_005263215.3 | XP_005263272.1 | 3, 4 | 546 | |||
| XM_017008602.1 | XP_016864091.1 | 3, 4 | 415 | |||
| GenBank mRNA | AB050785.1 | - | 3, 4 | 310 | ||
| AK123785.1 | - | 3, 4 | 549 | |||
| BC109029.2 | - | 3, 4 | 300 | |||
| BC109030.1 | - | 3, 4 | 299 | |||
| BC126899.1 | - | 3, 4 | 306 | |||
| BC128055.1 | - | 3, 4 | 310 | 40027832 | ||

ARHGAP10
Genomic Map