siRNA Details
| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024646.2 | NP_078922.1 | 9 | 1774 | ||
| XM_006710898.3 | XP_006710961.1 | 9 | 1661 | |||
| XM_017002336.1 | XP_016857825.1 | 9 | 1779 | |||
| GenBank mRNA | AB051517.1 | - | 9 | 1724 | ||
| AK023518.1 | - | 2 | 142 | |||
| AK056645.1 | - | 7 | 718 | |||
| AK095643.1 | - | 8 | 1414 | |||
| BC029832.1 | - | 7 | 788 | |||
| BX647643.1 | - | 7 | 947 | |||
| DQ646393.1 | - | 9 | 1793 | |||

ZYG11B
Genomic Map