siRNA Details
| Gene Symbol: THAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317776.1 | NP_001304705.1 | 6 | 2555 | ||
| NM_024672.5 | NP_078948.3 | 5 | 2489 | |||
| XM_005263220.2 | XP_005263277.1 | 5 | 2446 | |||
| XM_011532246.2 | XP_011530548.1 | 5 | 2279 | |||
| XM_017008603.1 | XP_016864092.1 | 5 | 3329 | |||
| XM_017008604.1 | XP_016864093.1 | 5 | 3382 | |||
| XM_017008605.1 | XP_016864094.1 | 5 | 2423 | |||
| XM_017008606.1 | XP_016864095.1 | 6 | 2547 | |||
| XM_017008607.1 | XP_016864096.1 | 5 | 2762 | |||
| XM_017008608.1 | XP_016864097.1 | 6 | 3401 | |||
| XM_017008609.1 | XP_016864098.1 | 6 | 2551 | |||
| XM_017008610.1 | XP_016864099.1 | 6 | 2538 | |||
| GenBank mRNA | AK026973.1 | - | 3 | 1819 | ||
| AK091412.1 | - | 5 | 2386 | |||
| AK225333.1 | - | 3 | 1826 | |||
| BC131722.1 | - | 6 | 2436 | |||
| BX648134.1 | - | 6 | 2709 | |||

THAP9
Genomic Map