siRNA Details
| Gene Symbol: ZNF665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024733.3 | NP_079009.3 | 4 | 1270 | ||
| XM_005259266.4 | XP_005259323.1 | 4 | 1304 | |||
| XM_011527325.2 | XP_011525627.1 | 4 | 1375 | |||
| XM_011527327.2 | XP_011525629.1 | 5 | 1679 | |||
| GenBank mRNA | AK291403.1 | - | 4 | 1260 | ||
| AL832507.1 | - | 4 | 1255 | |||

ZNF665
Genomic Map