siRNA Details
| Gene Symbol: C12orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024738.2 | NP_079014.1 | 2 | 347 | ||
| GenBank mRNA | AK025068.1 | - | 2 | 315 | ||
| AK222983.1 | - | 2 | 327 | |||
| BC019843.1 | - | 2 | 207 | 5112874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf49| Gene Symbol: C12orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024738.2 | NP_079014.1 | 2 | 347 | ||
| GenBank mRNA | AK025068.1 | - | 2 | 315 | ||
| AK222983.1 | - | 2 | 327 | |||
| BC019843.1 | - | 2 | 207 | 5112874 | ||