siRNA Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024756.2 | NP_079032.2 | 5 | 755 | ||
| XM_005270153.1 | XP_005270210.1 | 5 | 794 | |||
| XM_006717970.3 | XP_006718033.1 | 5 | 2734 | |||
| GenBank mRNA | AK023527.1 | - | 5 | 702 | ||
| AK304727.1 | - | 5 | 612 | |||
| AK308401.1 | - | 4 | 596 | |||
| BC064415.1 | - | 5 | 710 | |||
| BC072456.1 | - | 5 | 651 | |||
| BC094744.1 | - | 5 | 702 | 30343159 | ||

MMRN2
Genomic Map