siRNA Details
| Gene Symbol: KDM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145348.1 | NP_001138820.1 | 4 | 948 | ||
| NM_024773.2 | NP_079049.2 | 4 | 898 | |||
| XM_017023676.1 | XP_016879165.1 | 4 | 898 | |||
| XM_017023677.1 | XP_016879166.1 | 5 | 983 | |||
| XM_017023678.1 | XP_016879167.1 | 5 | 979 | |||
| XM_017023679.1 | XP_016879168.1 | 5 | 979 | |||
| XM_017023680.1 | XP_016879169.1 | 2 | 490 | |||
| GenBank mRNA | AK023860.1 | - | 4 | 889 | ||
| AK225066.1 | - | 4 | 855 | |||
| AK297166.1 | - | 4 | 948 | |||
| AK298410.1 | - | 1 | 18 | |||
| AK308499.1 | - | 2 | 212 | |||
| BC027911.1 | - | 4 | 877 | 5207043 | ||
| KU178785.1 | - | 3 | 716 | |||
| KU178786.1 | - | 3 | 716 | |||

KDM8
Genomic Map