siRNA Details
| Gene Symbol: TM4SF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024795.3 | NP_079071.2 | 2, 3 | 288 | ||
| XM_011511876.2 | XP_011510178.1 | 2, 3 | 194 | |||
| GenBank mRNA | AK026453.1 | - | 2, 3 | 287 | ||
| AY358671.1 | - | 2, 3 | 280 | |||
| BC035754.1 | - | 2, 3 | 275 | 5760060 | ||
| BC137256.1 | - | 2, 3 | 274 | 9021253 | ||
| BC137257.1 | - | 2, 3 | 274 | 9021254 | ||
| DA926038.1 | - | 2, 3 | 288 | |||

TM4SF20
Genomic Map