siRNA Details
| Gene Symbol: CAAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167575.1 | NP_001161047.1 | 3 | 815 | ||
| NM_024828.3 | NP_079104.3 | 3 | 661 | |||
| GenBank mRNA | AK023719.1 | - | 3 | 635 | ||
| AK300739.1 | - | 3 | 429 | |||
| AK301673.1 | - | 3 | 359 | |||
| BC071953.1 | - | 3 | 560 | 5431506 | ||

CAAP1
Genomic Map