siRNA Details
| Gene Symbol: ZNF212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012256.3 | NP_036388.2 | 3 | 590 | ||
| XM_005250055.2 | XP_005250112.2 | 3 | 588 | |||
| XM_011516581.2 | XP_011514883.2 | 3 | 578 | |||
| XM_011516582.1 | XP_011514884.1 | 3 | 510 | |||
| XM_017012646.1 | XP_016868135.1 | 3 | 591 | |||
| XM_017012647.1 | XP_016868136.1 | 4 | 544 | |||
| XM_017012648.1 | XP_016868137.1 | 3 | 833 | |||
| GenBank mRNA | AK074821.1 | - | 4 | 593 | ||
| AK315086.1 | - | 3 | 599 | |||
| BC022785.2 | - | 3 | 557 | 3849803 | ||
| U38864.1 | - | 1 | 41 | |||

ZNF212
Genomic Map