siRNA Details
| Gene Symbol: SPEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024867.3 | NP_079143.3 | 22 | 3277 | ||
| XM_005248376.3 | XP_005248433.1 | 22 | 3382 | |||
| XM_005248377.3 | XP_005248434.1 | 22 | 3367 | |||
| XM_011514135.2 | XP_011512437.1 | 22 | 3387 | |||
| XM_011514136.2 | XP_011512438.1 | 22 | 3386 | |||
| XM_011514137.2 | XP_011512439.1 | 22 | 3371 | |||
| XM_011514138.2 | XP_011512440.1 | 22 | 3363 | |||
| XM_011514139.2 | XP_011512441.1 | 21 | 3311 | |||
| XM_011514140.2 | XP_011512442.1 | 21 | 3121 | |||
| XM_011514141.2 | XP_011512443.1 | 23 | 3585 | |||
| XM_011514142.1 | XP_011512444.1 | 22 | 3358 | |||
| XM_017009880.1 | XP_016865369.1 | 21 | 3203 | |||
| XM_017009881.1 | XP_016865370.1 | 22 | 4241 | |||
| XM_017009882.1 | XP_016865371.1 | 22 | 3375 | |||
| GenBank mRNA | AB051557.1 | - | 12 | 1618 | ||

SPEF2
Genomic Map