siRNA Details
| Gene Symbol: FAM110D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024869.2 | NP_079145.2 | 2 | 252 | ||
| GenBank mRNA | AK024112.1 | - | 2 | 252 | ||
| AK290715.1 | - | 2 | 192 | |||
| AU119858.1 | - | 2 | 252 | |||
| BC025658.1 | - | 2 | 238 | 5198480 | ||

FAM110D
Genomic Map