siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 19, 20 | 3753 | ||
| XM_006711816.3 | XP_006711879.1 | 19, 20 | 3575 | |||
| XM_011544278.2 | XP_011542580.2 | 12, 13 | 1364 | |||
| XM_017002391.1 | XP_016857880.1 | 18, 19 | 3317 | |||
| GenBank mRNA | AK074078.1 | - | 14, 15 | 2858 | ||
| AK092353.1 | - | 13, 14 | 2149 | |||
| AK097229.1 | - | 6, 7 | 582 | |||
| AK307812.1 | - | 6, 7 | 599 | |||
| BC008300.1 | - | 14, 15 | 1663 | |||

PCNX2
Genomic Map