siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 21 | 4033 | ||
| XM_006711816.3 | XP_006711879.1 | 21 | 3855 | |||
| XM_011544278.2 | XP_011542580.2 | 14 | 1644 | |||
| XM_017002391.1 | XP_016857880.1 | 20 | 3597 | |||
| XM_017002392.1 | XP_016857881.1 | 8 | 883 | |||
| GenBank mRNA | AK055374.1 | - | 2 | 199 | ||
| AK074078.1 | - | 16 | 3138 | |||
| AK092353.1 | - | 15 | 2429 | |||
| AK097229.1 | - | 8 | 862 | |||
| AK307812.1 | - | 8 | 879 | |||
| BC008300.1 | - | 16 | 1943 | |||

PCNX2
Genomic Map