siRNA Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030613.3 | NP_085116.2 | 5 | 695 | ||
| GenBank mRNA | AK131367.1 | - | 5 | 674 | ||
| BC132779.1 | - | 3 | 468 | 40146801 | ||
| BC132781.1 | - | 3 | 493 | 40146803 | ||
| BC142989.1 | - | 5 | 664 | 8860436 | ||
| BC144213.1 | - | 3 | 468 | 9052733 | ||
| BC144216.1 | - | 4 | 579 | 9052736 | ||

ZFP2
Genomic Map