siRNA Details
| Gene Symbol: THOC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025075.3 | NP_079351.2 | 1, 2 | 148 | ||
| GenBank mRNA | AI391685.1 | - | 1, 2 | 128 | ||
| AK027098.1 | - | 1, 2 | 58 | |||
| AK225110.1 | - | 1, 2 | 36 | |||
| BC065012.1 | - | 1, 2 | 18 | 6167514 | ||
| CR457373.1 | - | 1, 2 | 15 | |||

THOC7
Genomic Map