siRNA Details
| Gene Symbol: CTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025099.5 | NP_079375.3 | 10 | 1805 | ||
| NR_046431.1 | 10 | 1700 | ||||
| XM_006721577.3 | XP_006721640.1 | 9 | 1660 | |||
| XM_006721578.3 | XP_006721641.1 | 10 | 1789 | |||
| XM_011524010.2 | XP_011522312.1 | 10 | 1684 | |||
| XM_011524011.2 | XP_011522313.1 | 9 | 1554 | |||
| GenBank mRNA | AK091077.1 | - | 10 | 1756 | ||
| AL831955.1 | - | 10 | 1647 | |||
| BC110373.1 | - | 2 | 172 | |||
| BC111783.1 | - | 10 | 1756 | 40054070 | ||
| DQ451688.1 | - | 10 | 1746 | |||
| FJ790670.1 | - | 10 | 1746 | |||

CTC1
Genomic Map