siRNA Details
Gene Symbol: CEP290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025114.3 | NP_079390.3 | 5, 6 | 651 | ||
XM_011538756.2 | XP_011537058.1 | 5, 6 | 651 | |||
XM_011538757.2 | XP_011537059.1 | 5, 6 | 459 | |||
XM_011538758.2 | XP_011537060.1 | 5, 6 | 651 | |||
XM_011538759.2 | XP_011537061.1 | 5, 6 | 651 | |||
XM_011538760.1 | XP_011537062.1 | 5, 6 | 651 | |||
XM_011538761.1 | XP_011537063.1 | 5, 6 | 651 | |||
XM_011538762.2 | XP_011537064.1 | 5, 6 | 651 | |||
XM_011538763.2 | XP_011537065.1 | 5, 6 | 651 | |||
XM_011538764.2 | XP_011537066.1 | 5, 6 | 651 | |||
XM_011538765.2 | XP_011537067.1 | 5, 6 | 651 | |||
XM_017019980.1 | XP_016875469.1 | 5, 6 | 651 | |||
XM_017019981.1 | XP_016875470.1 | 5, 6 | 651 | |||
XM_017019982.1 | XP_016875471.1 | 5, 6 | 651 | |||
XM_017019983.1 | XP_016875472.1 | 5, 6 | 651 | |||
GenBank mRNA | AF317887.1 | - | 3, 4 | 219 | ||
AK023677.1 | - | 5, 6 | 651 | |||
BC043398.1 | - | 5, 6 | 405 | |||
DQ109808.1 | - | 5, 6 | 651 |
