siRNA Details
| Gene Symbol: ZKSCAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242894.1 | NP_001229823.1 | 5 | 931 | ||
| NM_001242895.1 | NP_001229824.1 | 3 | 350 | |||
| NM_024493.3 | NP_077819.2 | 4 | 814 | |||
| XM_005249423.3 | XP_005249480.1 | 3 | 980 | |||
| XM_006715215.2 | XP_006715278.1 | 4 | 712 | |||
| XM_006715216.2 | XP_006715279.1 | 4 | 727 | |||
| XM_006715218.3 | XP_006715281.1 | 3 | 278 | |||
| XM_017011320.1 | XP_016866809.1 | 5 | 808 | |||
| GenBank mRNA | DN831452.1 | - | 3 | 251 | ||

ZKSCAN3
Genomic Map