siRNA Details
| Gene Symbol: WDR82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025222.3 | NP_079498.2 | 1 | 353 | ||
| XM_011534136.2 | XP_011532438.1 | 1 | 47 | |||
| GenBank mRNA | AK291380.1 | - | 1 | 109 | ||
| AY358264.1 | - | 1 | 88 | |||
| BX648628.1 | - | 1 | 95 | |||
| DB500464.1 | - | 1 | 353 | |||

WDR82
Genomic Map