siRNA Details
| Gene Symbol: PDCD1LG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025239.3 | NP_079515.2 | 4 | 807 | ||
| XM_005251600.3 | XP_005251657.1 | 4 | 914 | |||
| GenBank mRNA | AF329193.1 | - | 4 | 792 | ||
| AF344424.1 | - | 4 | 807 | |||
| AK001872.1 | - | 2 | 238 | |||
| AY254343.1 | - | 3 | 534 | |||
| BC074766.2 | - | 4 | 568 | 30915184 | ||
| BC113678.1 | - | 4 | 570 | 8322730 | ||
| BC113680.1 | - | 4 | 570 | 8322732 | ||
| BC143920.1 | - | 4 | 570 | |||
| BX951459.1 | - | 2 | 444 | |||
| DQ336699.1 | - | 3 | 501 | |||
| EF444806.1 | - | 4 | 654 | |||

PDCD1LG2
Genomic Map