siRNA Details
| Gene Symbol: KHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251874.1 | NP_001238803.1 | 5 | 1080 | ||
| NM_030568.4 | NP_085045.3 | 4 | 796 | |||
| GenBank mRNA | BC022080.2 | - | 4 | 766 | 4813061 | |
| BM725230.1 | - | 3 | 339 | |||

KHDC1
Genomic Map