siRNA Details
| Gene Symbol: CPTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029885.1 | NP_001025056.1 | 2 | 433 | ||
| XM_005244801.3 | XP_005244858.1 | 2 | 546 | |||
| XM_011542200.2 | XP_011540502.1 | 2 | 442 | |||
| GenBank mRNA | JN542538.1 | - | 1 | 18 | ||

CPTP
Genomic Map