siRNA Details
| Gene Symbol: ZNF451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257273.1 | NP_001244202.1 | 4 | 1513 | ||
| XM_011514463.2 | XP_011512765.1 | 3 | 1270 | |||
| GenBank mRNA | AB051489.1 | - | 1 | 1465 | ||
| AK302577.1 | - | 4 | 1358 | |||
| BC042450.1 | - | 2 | 1179 | |||
| BC058853.1 | - | 2 | 1179 | |||

ZNF451
Genomic Map