siRNA Details
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257273.1 | NP_001244202.1 | 4 | 477 | ||
XM_011514463.2 | XP_011512765.1 | 3 | 234 | |||
GenBank mRNA | AB051489.1 | - | 1 | 429 | ||
AK302577.1 | - | 4 | 322 | |||
BC042450.1 | - | 2 | 143 | |||
BC058853.1 | - | 2 | 143 |
