siRNA Details
| Gene Symbol: OR4A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005272.3 | NP_001005272.3 | 1 | 292 | ||
| GenBank mRNA | BC140750.1 | - | 1 | 292 | 9021618 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4A5| Gene Symbol: OR4A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005272.3 | NP_001005272.3 | 1 | 292 | ||
| GenBank mRNA | BC140750.1 | - | 1 | 292 | 9021618 | |